Merck
  • The serine/threonine phosphatase inhibitor, calyculin A, inhibits and dissociates macrophage responses to lipopolysaccharide.
The serine/threonine phosphatase inhibitor, calyculin A, inhibits and dissociates macrophage responses to lipopolysaccharide.
Journal of immunology (Baltimore, Md. : 1950) (1995-08-01)
S A Barber, P Y Perera, R McNally, S N Vogel
ABSTRACT
LPS-stimulated macrophages (M phi) produce inflammatory mediators that are largely responsible for the pathophysiology associated with septic shock. M phi respond to LPS with rapid protein phosphorylation and dephosphorylation on serine, threonine, and tyrosine residues. If these events are critical for the cellular response to LPS, the kinases and/or phosphatases involved may be vulnerable targets for pharmacologic intervention. Recent studies demonstrated that tyrosine kinase inhibitors block LPS-induced tyrosine phosphorylation of MAP kinases as well as TNF-alpha and IL-1 beta production. To investigate a role for serine/threonine phosphatases, we evaluated the effect of calyculin A, a potent serine/threonine phosphatase inhibitor, on LPS stimulation of murine M phi. Pretreatment of M phi with calyculin A inhibited LPS-induced expression of six immediate-early genes: TNF-alpha, IL-1 beta, IFN-beta, IP-10, IRF-1, and TNFR-2. Calyculin A added 1.5 h after LPS treatment greatly reduced accumulation of IP-10, IRF-1, and TNFR-2 mRNA, but not TNF-alpha, IL-1 beta, and IFN-beta mRNA. Calyculin A, in the absence or presence of LPS, resulted in sustained tyrosine phosphorylation of the MAP kinases. These findings suggest that an "early" serine/threonine phosphatase activity is essential for LPS stimulation of M phi and that the activation of MAP kinases is not sufficient for the induction of these immediate-early genes. The requirement for a "late" phosphatase activity for expression of a subset of LPS-inducible genes dissociates at least two regulatory pathways in LPS signal transduction.