Merck
  • Bactericidal effect of solar water disinfection under real sunlight conditions.
Bactericidal effect of solar water disinfection under real sunlight conditions.
Applied and environmental microbiology (2008-03-25)
M Boyle, C Sichel, P Fernández-Ibáñez, G B Arias-Quiroz, M Iriarte-Puña, A Mercado, E Ubomba-Jaswa, K G McGuigan
ABSTRACT
Batch solar disinfection (SODIS) inactivation kinetics are reported for suspensions in water of Campylobacter jejuni, Yersinia enterocolitica, enteropathogenic Escherichia coli, Staphylococcus epidermidis, and endospores of Bacillus subtilis, exposed to strong natural sunlight in Spain and Bolivia. The exposure time required for complete inactivation (at least 4-log-unit reduction and below the limit of detection, 17 CFU/ml) under conditions of strong natural sunlight (maximum global irradiance, approximately 1,050 W m(-2) +/- 10 W m(-2)) was as follows: C. jejuni, 20 min; S. epidermidis, 45 min; enteropathogenic E. coli, 90 min; Y. enterocolitica, 150 min. Following incomplete inactivation of B. subtilis endospores after the first day, reexposure of these samples on the following day found that 4% (standard error, 3%) of the endospores remained viable after a cumulative exposure time of 16 h of strong natural sunlight. SODIS is shown to be effective against the vegetative cells of a number of emerging waterborne pathogens; however, bacterial species which are spore forming may survive this intervention process.
Product Description
Millipore
Brucella broth base, suitable for microbiology, NutriSelect® Plus
Sigma-Aldrich
LB Broth (Miller), Highly-referenced nutrient-rich microbial growth powder medium, suitable for regular E.coli culture