Merck
  • The mode of inhibitory action by aphidicolin on eukaryotic DNA polymerase alpha.
The mode of inhibitory action by aphidicolin on eukaryotic DNA polymerase alpha.
European journal of biochemistry (1979-07-01)
M Oguro, C Suzuki-Hori, H Nagano, Y Mano, S Ikegami
PMID467434
ABSTRACT
The mode of action by aphidicolin on DNA polymerase alpha from the nuclear fraction of sea-urchin blastulae was studied. The inhibition of DNA polymerase alpha by aphidicolin was uncompetive with activated DNA and competitive with the four deoxynucleoside triphosphates using activated DNA as a template-primer. For truncated (residual or limited) DNA synthesis with only three deoxynucleoside triphosphates, aphidicolin inhibited the residual synthesis more strongly in the absence of dCTP than in the absence of each of the other three deoxynucleoside triphosphates. The inhibition was reversed with excess dCTP but not with the other three deoxynucleoside triphosphates. That is, aphidicolin inhibited DNA polymerase alpha by competing with dCTP with a Ki value of 0.5 microgram/ml and by not competing with the other three deoxynucleoside triphosphates. dTMP incorporation with the activated DNA was more sensitive to aphidicolin than dGMP or dTMP incorporation with poly(dC). (dG)12-18 or poly(dA) . (dT)12-18. Similar results were obtained for DNA polymerase alpha (B form) from mouse myeloma MOPC 104E.
Sigma-Aldrich
Aphidicolin, Ready Made Solution, from Nigrospora sphaerica