Merck
  • Asymmetric hydrogenation of alpha-primary and secondary amino ketones: efficient asymmetric syntheses of (-)-arbutamine and (-)-denopamine.
Asymmetric hydrogenation of alpha-primary and secondary amino ketones: efficient asymmetric syntheses of (-)-arbutamine and (-)-denopamine.
Chemistry (Weinheim an der Bergstrasse, Germany) (2007-06-27)
Gao Shang, Duan Liu, Scott E Allen, Qin Yang, Xumu Zhang
ABSTRACT
Two beta-receptor agonists (-)-denopamine and (-)-arbutamine were prepared in good yields and enantioselectivities by asymmetric hydrogenation of unprotected amino ketones for the first time by using Rh catalysts bearing electron-donating phosphine ligands. A series of alpha-primary and secondary amino ketones were synthesized and hydrogenated to produce various 1,2-amino alcohols in good yields and with good enantioselectivies. This Rh electron-donating phosphine-catalyzed asymmetric hyderogenation represents one of the most promising and convenient approaches towards the asymmetric synthesis of chiral amino alcohols.
Sigma-Aldrich
R(−)-Denopamine, ≥98% (HPLC), powder