Merck
  • Melatonin triggers p53Ser phosphorylation and prevents DNA damage accumulation.
Melatonin triggers p53Ser phosphorylation and prevents DNA damage accumulation.
Oncogene (2011-10-18)
R Santoro, M Marani, G Blandino, P Muti, S Strano
ABSTRACT
Several epidemiological studies have shown that high levels of melatonin, an indolic hormone secreted mainly by the pineal gland, reduce the risks of developing cancer, thus suggesting that melatonin triggers the activation of tumor-suppressor pathways that lead to the prevention of malignant transformation. This paper illustrates that melatonin induces phosphorylation of p53 at Ser-15 inhibiting cell proliferation and preventing DNA damage accumulation of both normal and transformed cells. This activity requires p53 and promyelocytic leukemia (PML) expression and efficient phosphorylation of p53 at Ser-15 residue. Melatonin-induced p53 phosphorylation at Ser-15 residue does not require ataxia telangiectasia-mutated activity, whereas it is severely impaired upon chemical inhibition of p38 mitogen-activated protein kinase activity. By and large, these findings imply that the activation of the p53 tumor-suppressor pathway is a critical mediator of melatonin and its anticancer effects. Therefore, it provides molecular insights into increasing observational evidence for the role that melatonin has in cancer prevention.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
SB 202190, ≥98% (HPLC)
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Millipore
Immobilon®-P PVDF Membrane, 10 sheets, 8 cm x 10 cm, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.