Merck
XIST RNA associates with specific regions of the inactive X chromatin.
The Journal of biological chemistry (2000-09-28)
S L Gilbert, J R Pehrson, P A Sharp
ABSTRACT
Microscopy studies have shown that XIST RNA colocalizes with the inactive X chromosome (Xi). However, the molecular basis for this colocalization is unknown. Here we provide two lines of evidence from chromatin immunoprecipitation experiments that XIST RNA physically associates with the Xi chromatin. First, XIST RNA can be co-precipitated by antiserum against macroH2A, a histone H2A variant enriched in the Xi. Second, XIST RNA can be co-precipitated by antisera that recognize unacetylated, but not acetylated, isoforms of histones H3 and H4. The specificity of XIST RNA association with hypoacetylated chromatin, together with the previous finding that hypoacetylated histone H4 is enriched at promoters of X-inactivated genes, raises the possibility that XIST RNA may contribute to the hypoacetylation of specific regions of the Xi so as to alter the expression of X-linked genes.