Merck
  • Gab2 Ablation Reverses the Stemness of HER2-Overexpressing Breast Cancer Cells.
Gab2 Ablation Reverses the Stemness of HER2-Overexpressing Breast Cancer Cells.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2018-10-17)
Peng Zhang, Yiying Chen, Miaomiao Gong, Zhumei Zhuang, Yueyue Wang, Lin Mu, Tianjiao Wang, Jinjin Pan, Yanan Liu, Jianming Xu, Rui Liang, Yuhui Yuan
ABSTRACT
HER2 has been implicated in mammary tumorigenesis as well as aggressive tumor growth and metastasis. Its overexpression is related to a poor prognosis and chemoresistance in breast cancer patients. Although Grb2-associated binding protein 2 (Gab2) is important in the development and progression of human cancer, its effects and mechanisms in HER2-overexpressing breast cancer are unclear. Clone formation and MTT assays were used to examine cell proliferation. To detect the effect of Gab2 on the stemness of breast cancer cells, we used flow cytometry, a sphere formation assay, real-time PCR, and western blot. An animal model was created to validate the effect of Gab2 on tumor growth in vivo. Tissue slides were analyzed by immunohistochemistry. Knockdown of Gab2 suppressed PI3K/AKT and MAPK/ERK pathway activity. Gab2 ablation also reduced the stemness of HER2-overexpressing breast cancer cells. In vivo, knockdown of Gab2 inhibited tumor growth. This study unveils a potential function of Gab2 in HER2-overexpressing breast cancer cells. Gab2 might be a potential target in the clinical therapy of HER2-overexpressing breast carcinoma.
Sigma-Aldrich
MISSION® esiRNA, targeting human GAB2