Merck
  • Filamin C Truncation Mutations Are Associated With Arrhythmogenic Dilated Cardiomyopathy and Changes in the Cell-Cell Adhesion Structures.
Filamin C Truncation Mutations Are Associated With Arrhythmogenic Dilated Cardiomyopathy and Changes in the Cell-Cell Adhesion Structures.
JACC. Clinical electrophysiology (2018-08-02)
Rene L Begay, Sharon L Graw, Gianfranco Sinagra, Angeliki Asimaki, Teisha J Rowland, Dobromir B Slavov, Katherine Gowan, Kenneth L Jones, Francesca Brun, Marco Merlo, Daniela Miani, Mary Sweet, Kalpana Devaraj, Eric P Wartchow, Marta Gigli, Ilaria Puggia, Ernesto E Salcedo, Deborah M Garrity, Amrut V Ambardekar, Peter Buttrick, T Brett Reece, Michael R Bristow, Jeffrey E Saffitz, Luisa Mestroni, Matthew R G Taylor
ABSTRACT
The purpose of this study was to assess the phenotype of Filamin C (FLNC) truncating variants in dilated cardiomyopathy (DCM) and understand the mechanism leading to an arrhythmogenic phenotype. Mutations in FLNC are known to lead to skeletal myopathies, which may have an associated cardiac component. Recently, the clinical spectrum of FLNC mutations has been recognized to include a cardiac-restricted presentation in the absence of skeletal muscle involvement. A population of 319 U.S. and European DCM cardiomyopathy families was evaluated using whole-exome and targeted next-generation sequencing. FLNC truncation probands were identified and evaluated by clinical examination, histology, transmission electron microscopy, and immunohistochemistry. A total of 13 individuals in 7 families (2.2%) were found to harbor 6 different FLNC truncation variants (2 stopgain, 1 frameshift, and 3 splicing). Of the 13 FLNC truncation carriers, 11 (85%) had either ventricular arrhythmias or sudden cardiac death, and 5 (38%) presented with evidence of right ventricular dilation. Pathology analysis of 2 explanted hearts from affected FLNC truncation carriers showed interstitial fibrosis in the right ventricle and epicardial fibrofatty infiltration in the left ventricle. Ultrastructural findings included occasional disarray of Z-discs within the sarcomere. Immunohistochemistry showed normal plakoglobin signal at cell-cell junctions, but decreased signals for desmoplakin and synapse-associated protein 97 in the myocardium and buccal mucosa. We found FLNC truncating variants, present in 2.2% of DCM families, to be associated with a cardiac-restricted arrhythmogenic DCM phenotype characterized by a high risk of life-threatening ventricular arrhythmias and a pathological cellular phenotype partially overlapping with arrhythmogenic right ventricular cardiomyopathy.
Sigma-Aldrich
Monoclonal Anti-Plakoglobin (Catenin γ) antibody produced in mouse, clone 15F11, ascites fluid, buffered aqueous solution