Merck
  • Trazodone alleviates both dyskinesia and psychosis in the parkinsonian marmoset model of Parkinson's disease.
Trazodone alleviates both dyskinesia and psychosis in the parkinsonian marmoset model of Parkinson's disease.
Journal of neural transmission (Vienna, Austria : 1996) (2017-12-17)
Adjia Hamadjida, Stephen G Nuara, Jim C Gourdon, Philippe Huot
ABSTRACT
Trazodone is a clinically available anti-depressant that exhibits affinity for serotonin 1A and 2A receptors, as well as for alpha-adrenoceptors, suggesting that it may be useful to treat L-3,4-dihydroxyphenylalanine (L-DOPA)-induced dyskinesia and psychosis that are encountered in advanced Parkinson's disease (PD). Here, we investigated the anti-dyskinetic and anti-psychotic effects of trazodone in the parkinsonian non-human primate. 6 common marmosets were rendered parkinsonian by administration of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP). Following repeated administration of L-DOPA to induce stable dyskinesia and psychosis-like behaviours (PLBs), trazodone (0.1, 1 and 10 mg/kg) or vehicle was administered in combination with L-DOPA and its effects on dyskinesia, PLBs and parkinsonism were determined. The addition of trazodone 10 mg/kg to L-DOPA reduced peak dose dyskinesia by ≈ 39% (P < 0.01) and peak dose PLBs by ≈ 17% (P < 0.01). However, parkinsonian disability was significantly worsened by trazodone 10 mg/kg (P < 0.05) and duration of anti-parkinsonian action was diminished by ≈ 21% (P < 0.05). Our results suggest that trazodone may be effective in alleviating L-DOPA-induced dyskinesia and psychosis in PD, but its deleterious effect on motor function is a concern and may limit its tolerability and usefulness in clinical settings.