Merck
  • ER-associated ubiquitin ligase HRD1 programs liver metabolism by targeting multiple metabolic enzymes.
ER-associated ubiquitin ligase HRD1 programs liver metabolism by targeting multiple metabolic enzymes.
Nature communications (2018-09-12)
Juncheng Wei, Yanzhi Yuan, Lu Chen, Yuanming Xu, Yuehui Zhang, Yajun Wang, Yanjie Yang, Clara Bien Peek, Lauren Diebold, Yi Yang, Beixue Gao, Chaozhi Jin, Johanna Melo-Cardenas, Navdeep S Chandel, Donna D Zhang, Hui Pan, Kezhong Zhang, Jian Wang, Fuchu He, Deyu Fang
ABSTRACT
The HMG-CoA reductase degradation protein 1 (HRD1) has been identified as a key enzyme for endoplasmic reticulum-associated degradation of misfolded proteins, but its organ-specific physiological functions remain largely undefined. Here we show that mice with HRD1 deletion specifically in the liver display increased energy expenditure and are resistant to HFD-induced obesity and liver steatosis and insulin resistance. Proteomic analysis identifies a HRD1 interactome, a large portion of which includes metabolic regulators. Loss of HRD1 results in elevated ENTPD5, CPT2, RMND1, and HSD17B4 protein levels and a consequent hyperactivation of both AMPK and AKT pathways. Genome-wide mRNA sequencing revealed that HRD1-deficiency reprograms liver metabolic gene expression profiles, including suppressing genes involved in glycogenesis and lipogenesis and upregulating genes involved in glycolysis and fatty acid oxidation. We propose HRD1 as a liver metabolic regulator and a potential drug target for obesity, fatty liver disease, and insulin resistance associated with the metabolic syndrome.
Sigma-Aldrich
Anti-SYVN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, HRP-conjugate, 1 mg/mL, Upstate®
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-ATP5D antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-HMGCR (internal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, HRP conjugate, Upstate®, from goat