Merck
  • Multiplicity generates diversity in the retinoic acid signalling pathways.
Multiplicity generates diversity in the retinoic acid signalling pathways.
Trends in biochemical sciences (1992-10-01)
M Leid, P Kastner, P Chambon
ABSTRACT
Complexity in the retinoid signalling system arises from a combination of several forms of retinoic acid, multiple cytoplasmic binding proteins and nuclear receptors, and the existence of polymorphic retinoic acid response elements. Additional diversity appears to be generated by heterodimeric interactions between two families of nuclear retinoic acid receptors, and between nuclear retinoic acid receptors and other members of the nuclear receptor superfamily. Thus, a complex array of combinatorial effects is beginning to emerge which may account for the pleiotropic effects of retinoids.
Sigma-Aldrich
RAR β-ligand binding domain (173-409) human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
RARβ, ligand binding domain (173-409), GST tagged human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
RAR, β human, recombinant, expressed in E. coli, ≥75% (SDS-PAGE)
Sigma-Aldrich
RAR, γ isoform, GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE)
Sigma-Aldrich
RAR, α human, recombinant, expressed in insect cells, ≥70% (SDS-PAGE)
Sigma-Aldrich
RARα-ligand binding domain (154-462) human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE)
Sigma-Aldrich
RARα, ligand binding domain (154-462), GST tagged human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
RAR γ-ligand binding domain (150-417) human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
RARγ, ligand binding domain (150-417), GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE)