Merck
  • Allele exchange at the EPSPS locus confers glyphosate tolerance in cassava.
Allele exchange at the EPSPS locus confers glyphosate tolerance in cassava.
Plant biotechnology journal (2017-12-10)
Aaron W Hummel, Raj Deepika Chauhan, Tomas Cermak, Andrew M Mutka, Anupama Vijayaraghavan, Adam Boyher, Colby G Starker, Rebecca Bart, Daniel F Voytas, Nigel J Taylor
ABSTRACT
Effective weed control can protect yields of cassava (Manihot esculenta) storage roots. Farmers could benefit from using herbicide with a tolerant cultivar. We applied traditional transgenesis and gene editing to generate robust glyphosate tolerance in cassava. By comparing promoters regulating expression of transformed 5-enolpyruvylshikimate-3-phosphate synthase (EPSPS) genes with various paired amino acid substitutions, we found that strong constitutive expression is required to achieve glyphosate tolerance during in vitro selection and in whole cassava plants. Using strategies that exploit homologous recombination (HR) and nonhomologous end-joining (NHEJ) DNA repair pathways, we precisely introduced the best-performing allele into the cassava genome, simultaneously creating a promoter swap and dual amino acid substitutions at the endogenous EPSPS locus. Primary EPSPS-edited plants were phenotypically normal, tolerant to high doses of glyphosate, with some free of detectable T-DNA integrations. Our methods demonstrate an editing strategy for creating glyphosate tolerance in crop plants and demonstrate the potential of gene editing for further improvement of cassava.
Sigma-Aldrich
Fluoroethylene carbonate, 99%
Pricing and availability is not currently available.