Merck
  • Assessment of factors affecting micropropagation and ex vitro acclimatization of Nyctanthes arbor-tristis L.
Assessment of factors affecting micropropagation and ex vitro acclimatization of Nyctanthes arbor-tristis L.
Acta biologica Hungarica (2011-03-11)
Anushi Arjumend Jahan, M Anis, I M Aref
ABSTRACT
Rapid differentiation of multiple shoots was observed in 94% of nodal explants of one year old Nyctanthes arbor-tristis L. plants. Shoot bud induction and multiplication took place on Murashige and Skoog (MS) medium supplemented with two cytokinins, i.e. Benzyladenine (BA) or Kinetin (Kn) either alone or in combination with different auxins, indole-3-butyric acid (IBA), indole-3-acetic acid (IAA) or α-naphthalene acetic acid (NAA). Between different media, pH levels and growth regulators tried, the optimum condition for maximum regenerative response was obtained on MS + Kn (2.5 μM) + N AA (0.5 μM) media at 5.8 pH, forming cultures with 23.26 ± 0.89 number of shoots and 6.36 ± 0.80 cm shoot length after 8 weeks of culture. Histological sections confirmed the formation of multiple buds from nodal explants. Rooting was achieved ex vitro by dipping the basal ends of microshoots in 200 μM IBA for 30 min followed by their transplantation in sterile soilrite. The plantlets with well-developed shoot and root system were successfully established in garden soil and grown outside in a greenhouse with a 80% survival rate.
Sigma-Aldrich
DL-Homophenylalanine, 98%
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
L-Homophenylalanine hydrochloride, 97%
Sigma-Aldrich
Murashige and Skoog Basal Medium, suitable for plant cell culture, with sucrose and agar
Sigma-Aldrich
Murashige and Skoog Basal Medium, powder, suitable for plant cell culture
Sigma-Aldrich
Murashige and Skoog Basal Medium, powder, suitable for plant cell culture, with Gamborg′s vitamins