Merck
  • Role of GA3, GA4 and uniconazole-P in controlling gravitropism and tension wood formation in Fraxinus mandshurica Rupr. var. japonica Maxim. seedlings.
Role of GA3, GA4 and uniconazole-P in controlling gravitropism and tension wood formation in Fraxinus mandshurica Rupr. var. japonica Maxim. seedlings.
Journal of integrative plant biology (2008-08-01)
Sha Jiang, Ke Xu, Yong-Zhou Wang, Yan-Ping Ren, Song Gu
ABSTRACT
GA(3) and GA(4) (gibberellins) play an important role in controlling gravitropism and tension wood formation in woody angiosperms. In order to improve our understanding of the role of GA(3) and GA(4) on xylem cell formation and the G-layer, we studied the effect of GA(3) and GA(4) and uniconazole-P, which is an inhibitor of GA biosynthesis, on tension wood formation by gravity in Fraxinus mandshurica Rupr. var. japonica Maxim. seedlings. Forty seedlings were divided into two groups; one group was placed upright and the other tilted. Each group was further divided into four sub-groups subjected to the following treatments: 3.43 x 10(-9) micromol acetone as control, 5.78 x 10(-8) micromol gibberellic acid (GA(3)), 6.21 x 10(-8) micromol GA(4), and 6.86 x 10(-8) micromol uniconazole-P. During the experimental period, GAs-treated seedlings exhibited negative gravitropism, whereas application of uniconazole-P inhibited negative gravitropic stem bending. GA(3) and GA(4) promoted wood fibers that possessed a gelatinous layer on the upper side, whereas uniconazole-P inhibited wood formation but did not inhibit the differentiation of the gelatinous layer in wood fibers on the upper side. These results suggest that: (i) both the formation of gelatinous fibers and the quantity of xylem production are important for the negative gravitropism in horizontally-positioned seedlings; (ii) GA(3) and GA(4) affect wood production more than differentiation of the gelatinous layer in wood fibers; G-layer development may be regulated by other hormones via the indirect-role of GA(3) and GA(4) in horizontally-positioned F. mandshurica seedlings rather than the direct effect of GAs; and (iii) the mechanism for upward wood stem bending is different to the newly developed shoot bending in reaction to gravity in this species.
Sigma-Aldrich
Gibberellin A4, suitable for plant cell culture, BioReagent, ≥90%
Sigma-Aldrich
Gibberellic acid, suitable for plant cell culture, BioReagent, ≥90% gibberellin A3 basis (of total gibberellins.)
Sigma-Aldrich
Gibberellic acid potassium salt, suitable for plant cell culture, BioReagent, ~95%, ≥50% total GA3 basis