Merck
  • Identification of spinal circuits involved in touch-evoked dynamic mechanical pain.
Identification of spinal circuits involved in touch-evoked dynamic mechanical pain.
Nature neuroscience (2017-04-25)
Longzhen Cheng, Bo Duan, Tianwen Huang, Yan Zhang, Yangyang Chen, Olivier Britz, Lidia Garcia-Campmany, Xiangyu Ren, Linh Vong, Bradford B Lowell, Martyn Goulding, Yun Wang, Qiufu Ma
ABSTRACT
Mechanical hypersensitivity is a debilitating symptom for millions of chronic pain patients. It exists in distinct forms, including brush-evoked dynamic and filament-evoked punctate hypersensitivities. We reduced dynamic mechanical hypersensitivity induced by nerve injury or inflammation in mice by ablating a group of adult spinal neurons defined by developmental co-expression of VGLUT3 and Lbx1 (VT3Lbx1 neurons): the mice lost brush-evoked nocifensive responses and conditional place aversion. Electrophysiological recordings show that VT3Lbx1 neurons form morphine-resistant polysynaptic pathways relaying inputs from low-threshold Aβ mechanoreceptors to lamina I output neurons. The subset of somatostatin-lineage neurons preserved in VT3Lbx1-neuron-ablated mice is largely sufficient to mediate morphine-sensitive and morphine-resistant forms of von Frey filament-evoked punctate mechanical hypersensitivity. Furthermore, acute silencing of VT3Lbx1 neurons attenuated pre-established dynamic mechanical hypersensitivity induced by nerve injury, suggesting that these neurons may be a cellular target for treating this form of neuropathic pain.
Sigma-Aldrich
Diphtheria Toxin from Corynebacterium diphtheriae, lyophilized powder, Product is in unnicked form
Sigma-Aldrich
(+)-Bicuculline, ≥97.0% (TLC)
Sigma-Aldrich
Anti-c-Fos Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Clozapine N-oxide
Sigma-Aldrich
Anti-Substance P Receptor antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Strychnine