Merck
  • Helicobacter suis binding to carbohydrates on human and porcine gastric mucins and glycolipids occurs via two modes.
Helicobacter suis binding to carbohydrates on human and porcine gastric mucins and glycolipids occurs via two modes.
Virulence (2018-04-12)
Médea Padra, Barbara Adamczyk, John Benktander, Bram Flahou, Emma C Skoog, János Tamás Padra, Annemieke Smet, Chunsheng Jin, Richard Ducatelle, Tore Samuelsson, Freddy Haesebrouck, Niclas G Karlsson, Susann Teneberg, Sara K Lindén
ABSTRACT
Helicobacter suis colonizes the stomach of most pigs and is the most prevalent non-Helicobacter pylori Helicobacter species found in the human stomach. In the human host, H. suis contributes to the development of chronic gastritis, peptic ulcer disease and MALT lymphoma, whereas in pigs it is associated with gastritis, decreased growth and ulcers. Here, we demonstrate that the level of H. pylori and H. suis binding to human and pig gastric mucins varies between individuals with species dependent specificity. The binding optimum of H. pylori is at neutral pH whereas that of H. suis has an acidic pH optimum, and the mucins that H. pylori bind to are different than those that H. suis bind to. Mass spectrometric analysis of mucin O-glycans from the porcine mucin showed that individual variation in binding is reflected by a difference in glycosylation; of 109 oligosaccharide structures identified, only 14 were present in all examined samples. H. suis binding to mucins correlated with glycans containing sulfate, sialic acid and terminal galactose. Among the glycolipids present in pig stomach, binding to lactotetraosylceramide (Galβ3GlcNAcβ3Galβ4Glcβ1Cer) was identified, and adhesion to Galβ3GlcNAcβ3Galβ4Glc at both acidic and neutral pH was confirmed using other glycoconjugates. Together with that H. suis bound to DNA (used as a proxy for acidic charge), we conclude that H. suis has two binding modes: one to glycans terminating with Galβ3GlcNAc, and one to negatively charged structures. Identification of the glycan structures H. suis interacts with can contribute to development of therapeutic strategies alternative to antibiotics.
Sigma-Aldrich
Monoclonal Anti-Mucin Gastric antibody produced in mouse, clone 45M1, ascites fluid
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine (TMB) Liquid Substrate System for ELISA, peroxidase substrate