Merck
  • Activation of p38 mitogen-activated protein kinase is required for tumor necrosis factor-alpha -supported proliferation of leukemia and lymphoma cell lines.
Activation of p38 mitogen-activated protein kinase is required for tumor necrosis factor-alpha -supported proliferation of leukemia and lymphoma cell lines.
The Journal of biological chemistry (2000-04-28)
R Y Liu, C Fan, G Liu, N E Olashaw, K S Zuckerman
ABSTRACT
To elucidate mechanisms of tumor necrosis factor alpha (TNF-alpha)-induced proliferation of a number of human leukemia and lymphoma cell lines, we examined the role of p38 mitogen-activated protein kinase (MAPK) in TNF-alpha signaling in Mo7e and Hut-78 cells. TNF-alpha-dependent p38 MAPK activation was detected in both Mo7e and Hut-78 cells and was blocked by the p38 MAPK inhibitor, SB203580. Ablation of p38 MAPK activity by SB203580 abrogated TNF-alpha-induced Mo7e cell proliferation and TNF-alpha-dependent autocrine growth of Hut-78. As we have shown previously that activation of the nuclear factor kappaB (NF-kappaB) is also required for TNF-alpha-induced Mo7e cell proliferation, the involvement of p38 MAPK in NF-kappaB activation was assessed. SB203580 did not affect TNF-alpha-signaled nuclear translocation and DNA-binding activity of NF-kappaB, and inhibition of NF-kappaB function did not affect TNF-alpha-induced p38 MAPK activation, indicating that these events are not dependent on each other. However, SB203580 depressed the expression of NF-kappaB-dependent genes, as monitored by a kappaB-driven reporter gene. Our findings demonstrate that activation of both p38 MAPK and NF-kappaB plays a critical role in TNF-alpha-mediated survival and proliferation of human leukemia and lymphoma cells, and p38 MAPK acts at least in part by facilitating the transcriptional activation function of NF-kappaB.
Sigma-Aldrich
Anti-p38 MAP Kinase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution