- WWC3 regulates the Wnt and Hippo pathways via Dishevelled proteins and large tumour suppressor 1, to suppress lung cancer invasion and metastasis.
WWC3 regulates the Wnt and Hippo pathways via Dishevelled proteins and large tumour suppressor 1, to suppress lung cancer invasion and metastasis.
The scaffolding protein WWC (WW and C2-domain containing) family is known to regulate cell proliferation and organ size via the Hippo signalling pathway. However, the expression level of WWC3 in human tumours and the mechanisms underlying its role in cellular signal transduction have not yet been reported. Herein, we explored the potential roles of WWC3 in lung cancer cells and the corresponding molecular mechanisms. We found low WWC3 expression in both lung cancer cell lines and lung cancer specimens, which was associated with low differentiation, advanced pTNM stage, positive lymph node metastasis, and poor prognosis in patients with lung cancer. Moreover, the overexpression of WWC3 inhibited the proliferation and invasiveness of lung cancer cells. These effects were mediated by the inhibition and stimulation of the Wnt and Hippo pathways, respectively, in vitro and in vivo. Specifically, WWC3 interacts with Dishevelled (Dvl) proteins, prevents casein kinase 1ϵ from phosphorylating Dvls, and inhibits β-catenin nuclear translocation to inhibit the Wnt pathway. Deleting the WW and C-terminal PDZ-binding domains of WWC3 abrogated these effects. Moreover, the interaction of WWC3 with Dvls reduced the interaction between WWC3 and large tumour suppressor 1 (LATS1), as well as decreasing LATS1 phosphorylation to increase the nuclear importation of yes-associated protein (YAP) and attenuate the Hippo pathway. Deleting the WW domain of WWC3 abrogated this effect. These findings demonstrate the molecular interplay between WWC3, Dvls, and LATS1, and reveal a link between the Wnt and Hippo pathways, which provides a potential target for clinical intervention in lung cancer. Copyright © 2017 Pathological Society of Great Britain and Ireland. Published by John Wiley & Sons, Ltd.