Merck
SCYL1 does not regulate REST expression and turnover.
PloS one (2017-06-02)
Sebastien Gingras, Emin Kuliyev, Stéphane Pelletier
ABSTRACT
A recent study identified SCYL1 as one of the components of the oncogenic STP axis, which promotes triple-negative breast cancer by regulating degradation of the REST tumor suppressor. Contrary to the findings of that study, herein we show by using 3 distinct genetic approaches that SCYL1 does not regulate REST turnover. Specifically, REST protein levels and turnover were identical in Scyl1+/+ and Scyl1-/- mouse embryonic fibroblasts. Similarly, targeted inactivation of SCYL1 in Hek293T cells by using CRIPSR-Cas9 technology did not affect REST steady-state level and turnover. Furthermore, RNA interference-mediated depletion of SCYL1 in Hek293T or MDA-MB-231 cells did not alter REST steady-state level and turnover. Together, our findings indicate that SCYL1 does not contribute to REST turnover and thus do not support a previous study suggesting a role for SCYL1 in mediating REST degradation.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-SCYL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-REST Antibody, Upstate®, from rabbit