Merck
  • Human remyelination promoting antibody inhibits apoptotic signaling and differentiation through Lyn kinase in primary rat oligodendrocytes.
Human remyelination promoting antibody inhibits apoptotic signaling and differentiation through Lyn kinase in primary rat oligodendrocytes.
Glia (2010-07-21)
J Watzlawik, E Holicky, D D Edberg, D L Marks, A E Warrington, B R Wright, R E Pagano, M Rodriguez
ABSTRACT
Human remyelination promoting IgM mAbs target oligodendrocytes (OLs) and function in animal models of multiple sclerosis (MS). However, their mechanism of action is unknown. This study seeks to identify the cellular mechanism of action of a recombinant human IgM on OL survival. Binding of rHIgM22 to the surface of rat OLs was studied by co-localization with various markers. RHIgM22-mediated effects on apoptotic signaling in OLs, differentiation markers, and signaling molecules were detected by Western blotting and immunoprecipitation. RHIgM22 co-localized with integrin β3 but not other integrin β-chains in OLs. Downstream of integrin β3 we identified Src family kinase (SFK) Lyn as a key player of rHIgM22-mediated actions in OLs. Lyn immunoprecipitated in a complex together with integrin αvβ3 and PDGFαR. Lyn expression was 9-fold up-regulated and Lyn activation was 3-fold higher inrHIgM22-treated OL cultures compared with controls. RHIgM22 inhibited apoptotic signaling by greater than 10-fold reduction of caspase-3 and capsase-9 cleavage and reduced by 4-fold expression of differentiation markers MBP and MOG in OLs. SFK inhibitors PP2 and SU6656 inhibited Lyn activity and restored caspase-cleavage in OLs. A human IgM that did not promote remyelination and medium wereused as controls. rHIgM22 prevented apoptotic signaling andinhibited OL differentiation by Lyn implying thatIgM-mediated remyelination is due toprotection of OPC and OLs rather than promotion of OPC differentiation.
Sigma-Aldrich
PP2, InSolution, ≥95%, 10 mM, reversible ATP-competitive inhibitor of the Src family of protein tyrosine kinases
Sigma-Aldrich
FGF-2 / basic FGF Protein, Human recombinant, The FGF-2 / basic FGF Protein, Human recombinant is available in a 25 µg format.
Sigma-Aldrich
SU6656, InSolution, ≥95%, inhibitor of Src-family protein tyrosine kinases
Sigma-Aldrich
Anti-Integrin β5 Antibody, CT/cytosolic, serum, Chemicon®