Skip to Content
Merck
Vigabatrin European Pharmacopoeia (EP) Reference Standard

Vigabatrin

Vigabatrin European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Vigabatrin, (±)-Vigabatrin, (R,S)-4-Amino-5-hexenoic acid, (±)-γ-Vinyl-GABA, (±)-4-Aminohexenoic acid
Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade