
Tutin solution

Synonym(s):
(1aS,1bR,2S,2′R,5R,6S,6aR,7aR,8R)-Hexahydro-1b,6-dihydroxy-6a-methyl-8-(1-methylethenyl)spiro[2,5-methano-7H-oxireno[3,4]cyclopent[1,2-d]oxepin-7,2′-oxiran]-3(2H)-one solution
Empirical Formula (Hill Notation):
C15H18O6
CAS Number:
Molecular Weight:
294.30
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1