Skip to Content
Merck
TSKgel® QC-PAK GFC 200 HPLC Column phase diol, L × I.D. 15 cm × 7.8 mm, 5 μm particle size

TSKgel® QC-PAK GFC 200 (5 µm) HPLC Column

TSKgel® QC-PAK GFC 200 HPLC Column phase diol, L × I.D. 15 cm × 7.8 mm, 5 μm particle size
Synonym(s):
TSKgel® SWXL Columns

Products

Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1