![TSKgel® CM-2SW HPLC Column phase carboxymethyl, L × I.D. 25 cm × 4.6 mm, 5 μm particle size](/deepweb/assets/sigmaaldrich/product/images/741/828/380b4b21-7ea0-405a-8e67-0fe7fe44eecc/640/380b4b21-7ea0-405a-8e67-0fe7fe44eecc.jpg)
TSKgel® CM-2SW (5 µm) HPLC Columns
![TSKgel® CM-2SW HPLC Column phase carboxymethyl, L × I.D. 25 cm × 4.6 mm, 5 μm particle size](/deepweb/assets/sigmaaldrich/product/images/741/828/380b4b21-7ea0-405a-8e67-0fe7fe44eecc/640/380b4b21-7ea0-405a-8e67-0fe7fe44eecc.jpg)
Synonym(s):
TSKgel® CM-SW Columns
Products
Product Category
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing