Skip to Content
Merck
TSKgel® BioAssist® S HPLC Column phase sulfonic acid, L × I.D. 5 cm × 4.6 mm, 7 μm particle size, PEEK hardware

TSKgel® BioAssist® S (7 µm) HPLC Columns

TSKgel® BioAssist® S HPLC Column phase sulfonic acid, L × I.D. 5 cm × 4.6 mm, 7 μm particle size, PEEK hardware
Synonym(s):
TSKgel® BioAssist® S HPLC Column

Products

Product Category
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1