Skip to Content
Merck
Tris[2-(2-methoxyethoxy)ethyl]amine 95%

Tris[2-(2-methoxyethoxy)ethyl]amine

Tris[2-(2-methoxyethoxy)ethyl]amine 95%
Synonym(s):
TDA-1, Tris(3,6-dioxaheptyl)amine
Linear Formula:
(CH3OCH2CH2OCH2CH2)3N
CAS Number:
Molecular Weight:
323.43
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing