![Trimyristin European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/202/197/991bbfe1-6bd3-4c7a-91d9-723cbaa40830/640/991bbfe1-6bd3-4c7a-91d9-723cbaa40830.png)
Trimyristin
![Trimyristin European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/202/197/991bbfe1-6bd3-4c7a-91d9-723cbaa40830/640/991bbfe1-6bd3-4c7a-91d9-723cbaa40830.png)
Synonym(s):
Glyceryl trimyristate, 1,2,3-Tritetradecanoylglycerol, Glycerol trimyristate, Trimyristin
Linear Formula:
[CH3(CH2)12COOCH2]2CHOCO(CH2)12CH3
CAS Number:
Molecular Weight:
723.16
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing