Skip to Content
Merck
(Trimethylsilyl)methyl isocyanide 97%

(Trimethylsilyl)methyl isocyanide

(Trimethylsilyl)methyl isocyanide 97%
Synonym(s):
(Isocyanomethyl)trimethylsilane
Linear Formula:
(CH3)3SiCH2NC
CAS Number:
Molecular Weight:
113.23

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing