Skip to Content
Merck
Tridecanoic acid analytical standard

Tridecanoic acid

Tridecanoic acid analytical standard
Linear Formula:
CH3(CH2)11CO2H
CAS Number:
Molecular Weight:
214.34
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing