Tribulosin
Synonym(s):
(3β,5α,25S)-Spirostan-3-yl 6-deoxy-α-L-mannopyranosyl-(1→2)-O-[O-β-D-xylopyranosyl-(1→2)-O-[β-D-xylopyranosyl-(1→3)]-β-D-glucopyranosyl-(1→4)]-β-D-galactopyranoside
Empirical Formula (Hill Notation):
C55H90O25
CAS Number:
Molecular Weight:
1151.29
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing