![Triamcinolone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/468/980/188b621d-b4f8-4ee7-abbb-bdb9e3798464/640/188b621d-b4f8-4ee7-abbb-bdb9e3798464.png)
Triamcinolone
![Triamcinolone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/468/980/188b621d-b4f8-4ee7-abbb-bdb9e3798464/640/188b621d-b4f8-4ee7-abbb-bdb9e3798464.png)
Synonym(s):
9α-Fluoro-11β,16α,17,21-tetrahydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-11β,16α,17,21-tetrahydroxypregna-1,4-diene-3,20-dione, Fluoxyprednisolone, Triamcinolone
Empirical Formula (Hill Notation):
C21H27FO6
CAS Number:
Molecular Weight:
394.43
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4