
TOYOPEARL GigaCap® DEAE-650 MiniChrom Column

Synonym(s):
MiniChrom
Products
Product Category
Manufacturer
Product Line
Analyte Suitability
Separation Technique
ID (mm)
Length (cm)
Volume (ml)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1