![Tetraethylene glycol 99%](/deepweb/assets/sigmaaldrich/product/structures/362/010/20e6723f-b37d-4f46-bfd5-c5be99f91146/640/20e6723f-b37d-4f46-bfd5-c5be99f91146.png)
Tetraethylene glycol
![Tetraethylene glycol 99%](/deepweb/assets/sigmaaldrich/product/structures/362/010/20e6723f-b37d-4f46-bfd5-c5be99f91146/640/20e6723f-b37d-4f46-bfd5-c5be99f91146.png)
Synonym(s):
Bis[2-(2-hydroxyethoxy)ethyl] ether, Tetra(ethylene glycol), Tetraglycol, Tetraethylene glycol
Linear Formula:
HO(CH2CH2O)4H
CAS Number:
Molecular Weight:
194.23
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Polymer Composition
Polymer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing