![tert-Butyllithium solution 1.6-3.2 M in heptane](/deepweb/assets/sigmaaldrich/product/structures/920/459/22ed7817-5ae4-4c65-b7a1-a19b9c4eb07f/640/22ed7817-5ae4-4c65-b7a1-a19b9c4eb07f.png)
tert-Butyllithium solution
![tert-Butyllithium solution 1.6-3.2 M in heptane](/deepweb/assets/sigmaaldrich/product/structures/920/459/22ed7817-5ae4-4c65-b7a1-a19b9c4eb07f/640/22ed7817-5ae4-4c65-b7a1-a19b9c4eb07f.png)
Synonym(s):
Lithium-2-methyl-2-propanide, t-BuLi
Linear Formula:
(CH3)3CLi
CAS Number:
Molecular Weight:
64.06
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing