Skip to Content
Merck
Terbutaline sulfate British Pharmacopoeia (BP) Reference Standard

Terbutaline Sulfate

Terbutaline sulfate British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Terbutaline hemisulfate salt, 2-t-Butylamino-1-(3,5-dihydroxyphenyl)ethanol, 5-[2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-1,3-benzenediol hemisulfate salt
Linear Formula:
C12H19NO3 · 1/2H2SO4
CAS Number:
Molecular Weight:
274.32
EC Number:

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade