Skip to Content
Merck
Temozolomide Related Compound B United States Pharmacopeia (USP) Reference Standard

Temozolomide Related Compound B

Temozolomide Related Compound B United States Pharmacopeia (USP) Reference Standard
Synonym(s):
3-Methyl-1,1-diphenylurea, N-Methyl-N′,N′-diphenylurea
Linear Formula:
(C6H5)2NCONHCH3
CAS Number:
Molecular Weight:
226.27

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1