Tanshinone IIA
Synonym(s):
1,6,6-Trimethyl-6,7,8,9-tetrahydrophenanthro[1,2-b]furan-10,11-dione, Dan Shen ketone
Empirical Formula (Hill Notation):
C19H18O3
CAS Number:
Molecular Weight:
294.34
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade