Skip to Content
Merck
Sudan I analytical standard

Sudan I

Sudan I analytical standard
Synonym(s):
1-Phenylazo-2-naphthol, Solvent Yellow 14
Linear Formula:
C6H5N=NC10H6OH
CAS Number:
Molecular Weight:
248.28
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing