Stigmasterol
Synonym(s):
3β-Hydroxy-24-ethyl-5,22-cholestadiene, 5,22-Stigmastadien-3β-ol, Stigmasterin
Empirical Formula (Hill Notation):
C29H48O
CAS Number:
Molecular Weight:
412.69
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing