Skip to Content
Merck
Spectinomycin dihydrochloride pentahydrate VETRANAL®, analytical standard

Spectinomycin dihydrochloride pentahydrate

Spectinomycin dihydrochloride pentahydrate VETRANAL®, analytical standard
Empirical Formula (Hill Notation):
C14H24N2O7 · 2HCl · 5H2O
CAS Number:
Molecular Weight:
495.35
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Activity Spectrum
Format
Quality Segment
Application
Microorganism Suitability
Mode of Action
Reference Material Type
Special Grade
Sterilization Method
Technique