Sodium thiocyanate solution
Synonym(s):
Sodium rhodanide solution
Empirical Formula (Hill Notation):
CNNaS
CAS Number:
Molecular Weight:
81.07
Products
Product Category
Brand
Formula Weight
pH Value
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing