Sodium biphenyl complex
Synonym(s):
Biphenyl sodium complex solution, Sodium biphenyl complex
Linear Formula:
C6H5C6H5 · Na
CAS Number:
Molecular Weight:
177.20
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Package Size
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing