![Sodium Alginate Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/379/057/9815601d-b06d-4c69-9477-547fe68db45d/640/9815601d-b06d-4c69-9477-547fe68db45d.jpg)
Sodium alginate
![Sodium Alginate Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/379/057/9815601d-b06d-4c69-9477-547fe68db45d/640/9815601d-b06d-4c69-9477-547fe68db45d.jpg)
Synonym(s):
Alginic acid sodium salt from brown algae, Algin, Sodium alginate, Alginic acid sodium salt
CAS Number:
Products
Product Category
Brand
Biological Source
Agency
Format
Quality Segment
Application
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing