Skip to Content
Merck
Silychristin Milk Thistle component, ≥95% (HPLC)

Silychristin

Silychristin Milk Thistle component, ≥95% (HPLC)
Synonym(s):
Silymarin II, Silychristin
Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing