![Seneciphylline phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/192/532/02bec92d-c301-4ac4-b273-112c1bc8b074/640/02bec92d-c301-4ac4-b273-112c1bc8b074.png)
Seneciphylline
![Seneciphylline phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/192/532/02bec92d-c301-4ac4-b273-112c1bc8b074/640/02bec92d-c301-4ac4-b273-112c1bc8b074.png)
Synonym(s):
α-Longilobin, 3,19-Didehydro-12-hydroxysenecionan-11,16-dione, Jacodine
Empirical Formula (Hill Notation):
C18H23NO5
CAS Number:
Molecular Weight:
333.38
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing