Skip to Content
Merck
(S)-(+)-Pantolactone 97%

(S)-(+)-Pantolactone

(S)-(+)-Pantolactone 97%
Synonym(s):
(S)-(+)-α-Hydroxy-β,β-dimethyl-γ-butyrolactone, (S)-(+)-β,β-Dimethyl-α-hydroxy-γ-butyrolactone, (S)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, L-Pantolactone
Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing