(S)-Bupivacaine
Synonym(s):
(2S)-1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide
Empirical Formula (Hill Notation):
C18H28N2O
CAS Number:
Molecular Weight:
288.43
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing