![Sarafloxacin hydrochloride hydrate VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/356/711/9f91e83f-ef8b-4f11-af82-f582f0db1d5b/640/9f91e83f-ef8b-4f11-af82-f582f0db1d5b.png)
Sarafloxacin hydrochloride hydrate
![Sarafloxacin hydrochloride hydrate VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/356/711/9f91e83f-ef8b-4f11-af82-f582f0db1d5b/640/9f91e83f-ef8b-4f11-af82-f582f0db1d5b.png)
Empirical Formula (Hill Notation):
C20H17F2N3O3 · HCl · xH2O
CAS Number:
Molecular Weight:
421.83 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing