Skip to Content
Merck
Salbutamol Impurity D Pharmaceutical Secondary Standard; Certified Reference Material

Salbutamol Impurity D

Salbutamol Impurity D Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
5-[2-(tert-Butylamino)-1-hydroxyethyl]-2-hydroxybenzaldehyde hemisulfate salt, 5-{2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl}-2-hydroxybenzaldehyde hemisulfate salt
Linear Formula:
(C13H19NO3)2 · H2SO4
CAS Number:
Molecular Weight:
572.67

Products

Brand
Formula Weight
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1