Saccharin calcium
Synonym(s):
Saccharin hemicalcium salt, 2,3-Dihydro-3-oxobenzisosulfonazole, o-Sulfobenzimide
Empirical Formula (Hill Notation):
C14H8CaN2O6S2 · 3.5H2O
CAS Number:
Molecular Weight:
467.48
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing